n-Octacosane ≥99%

Supplier: BeanTown Chemical

Synonyms: N-octacosane

136880-10G 136880-250G 136880-50G
BT136880-10GEA 23 USD
BT136880-10G BT136880-250G BT136880-50G
n-Octacosane ≥99%
n-Octacosane

CAS: 630-02-4; EC No: 211-125-7; MDL No: MFCD00009355 Crystalline/Powder; Linear Formula: CH3(CH2)26CH3; Molecular Formula: C28H58; MW: 394.77 Melting Point: 57-62°; Boiling Point: 440°; Flash point: 227°C (441°F)


  • Crystalline/Powder

Formula: H₃C(CH₂)₂₆CH₃
MW: 394.77 g/mol
Boiling Pt: 432
Melting Pt: 57…62 °C
Flash Pt: 227 °C (441 °F)
MDL Number: MFCD00009355
CAS Number: 630-02-4
EINECS: 211-125-7

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR