1-Heptanesulfonic acid sodium salt ≥99% (dry weight)

Supplier: BeanTown Chemical

Synonyms: Sodium heptane-1-sulfonate, Sodium 1-heptanesulfonate, Sodium heptane-1-sulphonate, Sodium 1-heptanesulphonate

136575-100G 136575-25G 136575-5G
BT136575-100GEA 424.08 USD
BT136575-100G BT136575-25G BT136575-5G
1-Heptanesulfonic acid sodium salt ≥99% (dry weight)
1-Heptanesulphonic acid sodium salt

CAS: 22767-50-6; EC No: 245-210-5; MDL No: MFCD00007543 Powder; Linear Formula: CH3(CH2)6SO3Na; Molecular Formula: C7H15NaO3S; MW: 202.24 Melting Point: 300°


  • Powder

Formula: H₃C(CH₂)₅CH₂SO₃Na
MW: 202.25 g/mol
Melting Pt: >300 °C
MDL Number: MFCD00007543
CAS Number: 22767-50-6
EINECS: 245-210-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=95c09a5691140bab7446d88919f01593e8f0b9b2&vid=7c237cde5db01d0567ff19964ca3287434747480&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR