Niobium ≥99.5% (trace metals basis), powder -60 mesh

Supplier: BeanTown Chemical

135895-100G 135895-25G 135895-500G
BT135895-100GEA 114.81 USD
BT135895-100G BT135895-25G BT135895-500G
Niobium ≥99.5% (trace metals basis), powder -60 mesh
Niobium

CAS: 7440-03-1; EC No: 231-113-5; MDL No: MFCD00011126 Powder; Molecular Formula: Nb ; MW: 92.91 Melting Point: 2468°; Boiling Point: 4742° Density (g/mL): 8.57


  • Powder

Formula: Nb
MW: 92.91 g/mol
Boiling Pt: 3700
Melting Pt: 2468
Density: 8.57 g/mL
MDL Number: MFCD00011126
CAS Number: 7440-03-1
EINECS: 231-113-5
UN: 3089
ADR: 4.1 II

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR