Copper(II) sulfide, anhydrous ≥99.98%
Supplier: BeanTown Chemical
Synonyms:
Copper sulfide, Cupric sulfide, Copper sulphide, Cupric sulphide, Copper(II)sulfide, Copper(II)sulphide
ampouled under argon
135890-10G
135890-50G
BT135890-10GEA
220
USD
BT135890-10G
BT135890-50G
Copper(II) sulfide, anhydrous ≥99.98%
Copper (II) sulphide
CAS: 1317-40-4; EC No: 215-271-2; MDL No: MFCD00016066
Powder; Molecular Formula: CuS ; MW: 95.60
Melting Point: 220° (decomposes)
Density (g/mL): 4.6
Air Sensitive, Moisture Sensitive
- Air Sensitive, Moisture Sensitive
- Powder
Formula:
CuS Melting Pt: 220 °C (dec.) Density: 4.6 g/mL |
MDL Number:
MFCD00016066 CAS Number: 1317-40-4 EINECS: 215-271-2 |
Specification Test Results
Assay | 99.98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...