3-Methylxanthine ≥98%
Supplier: BeanTown Chemical
Synonyms:
3,7-Dihydro-3-methyl-1H-purine-2,6-dione, 2,6-Dihydroxy-3-methylpurine
135695-1G
135695-250MG
BT135695-1GEA
219
USD
BT135695-1G
BT135695-250MG
3-Methylxanthine ≥98%
3-Methylxanthine
CAS: 1076-22-8; EC No: 214-058-1; MDL No: MFCD00005580; RTECS: ZD8750000
Powder; Molecular Formula: C6H6N4O2; MW: 166.14
Melting Point:
- RTECS: ZD8750000
- Powder
Formula:
C₆H₆N₄O₂ MW: 166.14 g/mol Melting Pt: >300 °C |
MDL Number:
MFCD00005580 CAS Number: 1076-22-8 EINECS: 214-058-1 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...