D-(+)-Maltose monohydrate ≥95%

Supplier: BeanTown Chemical

Synonyms: Maltobiose hydrate, Malt Sugar, beta-Maltose monohydrate, Maltose monohydrate, (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal hydrate, 4-O-α-D-Glucopyranosyl-D-glucose hydrate

135120-100G 135120-500G
BT135120-100GEA 28.43 USD
BT135120-100G BT135120-500G
D-(+)-Maltose monohydrate ≥95%
D(+)-Maltose monohydrate

CAS: 6363-53-7; EC No: 200-716-5; MDL No: MFCD00149343; RTECS: OO5250000 Powder; Molecular Formula: C12H22O11·H2O; MW: 360.31 Optical Rotation: [α]20/D +130±2°, c = 4% in water


  • RTECS: OO5250000
  • Optical rotation: [α]20/D +130±2°, c = 4% in water
  • Powder

Formula: C₁₂H₂₂O₁₁·1H₂O
MW: 360.32 g/mol
MDL Number: MFCD00149343
CAS Number: 6363-53-7
EINECS: 200-716-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f861d7adf53134a0c485b5942d3be34c4d0b571&vid=16e5c4425a7a8e158a4200470a45483c7ff5781e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR