Iron ≥99.99% (trace metals basis), rods Ø 5 mm
Supplier: BeanTown Chemical
134970-150MM
134970-50MM
BT134970-150MMEA
389
USD
BT134970-150MM
BT134970-50MM
Iron ≥99.99% (trace metals basis), rods Ø 5 mm
Iron
CAS: 7439-89-6; EC No: 231-096-4; MDL No: MFCD00010999
Solid; Molecular Formula: Fe; MW: 55.85
Melting Point: 1535°; Boiling Point: 2750°
Density (g/mL): 7.86
- Solid
Formula:
Fe MW: 55.85 g/mol Boiling Pt: 3000 Melting Pt: 1535 Density: 7.86 g/mL |
MDL Number:
MFCD00010999 CAS Number: 7439-89-6 EINECS: 231-096-4 Merck Index: 13,05109 |
Specification Test Results
Assay | 99.99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...