2,2-Bis(4-glycidyloxyphenyl)propane

Supplier: BeanTown Chemical

Synonyms: Bisphenol A Diglycidyl Ether, 2,2-Bis(4-hydroxyphenyl)propane Diglycidyl Ether, DER 332, BADGE, D.E.R. 332 epoxy resin

134745-100G 134745-25G 134745-500G
BT134745-100GEA 72.02 USD
BT134745-100G BT134745-25G BT134745-500G
2,2-Bis(4-glycidyloxyphenyl)propane
2,2-Bis(4-glycidyloxyphenyl)propane

CAS: 1675-54-3; EC No: 216-823-5; MDL No: MFCD00080480; RTECS: TX3800000 Liquid; Molecular Formula: C21H24O4; MW: 340.41 Density (g/mL): 1.16


  • RTECS: TX3800000
  • Liquid

Formula: C₂₁H₂₄O₄
MW: 340.42 g/mol
Density: 1.16 g/mL
MDL Number: MFCD00080480
CAS Number: 1675-54-3
EINECS: 216-823-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR