Platinum : rhodium (90:10) (w/w), wire 0.127 mm dia. for ISA type R standard grade thermocouple
Supplier: BeanTown Chemical
134500-1M
134500-5M
BT134500-1MEA
148
USD
BT134500-1M
BT134500-5M
Platinum : rhodium (90:10) (w/w), wire 0.127 mm dia. for ISA type R standard grade thermocouple
Platinum : rhodium
MDL No: MFCD00198186
Solid; Formula: Pt:Rh; 90:10 wt%
- Solid