Citric acid, anhydrous ≥99.5% ACS

Supplier: BeanTown Chemical

Synonyms: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid

134485-100G 134485-2KG 134485-500G
BT134485-100GEA 25 USD
BT134485-100G BT134485-2KG BT134485-500G
Citric acid, anhydrous ≥99.5% ACS
Citric acid

CAS: 77-92-9; EC No: 201-069-1; MDL No: MFCD00011669; RTECS: GE7350000 Crystalline; Molecular Formula: C6H8O7; MW: 192.13 Melting Point: 153-159° Hygroscopic


  • RTECS: GE7350000
  • Hygroscopic
  • Crystalline

Formula: HOC(COOH)(CH₂COOH)₂
MW: 192.13 g/mol
Boiling Pt: 310
Melting Pt: 153…159 °C
MDL Number: MFCD00011669
CAS Number: 77-92-9
EINECS: 201-069-1
Merck Index: 13,02350
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d60f5a9b8e6c6dfb7eda04e1a1d5d6d971f92444&vid=caf4f9effc7fd22b942f7f92be770b06e40637a4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.5%
Insoluble matter ≤0.005%
Chloride (Cl) ≤0.001%
Oxalate (C2O4) ≤0.05%
Phosphate (PO4) ≤0.001%
Sulfate (SO4) ≤0.002%
Iron (Fe) ≤3ppm
Lead (Pb) ≤2ppm
Residue after ignition ≤0.02%
Substances carbonizable by hot sulfuric acid Passes test
Tridodecylamine ≤0.1mg/kg
Water ≤0.5%
Clarity and color of solution Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR