Silicic acid hydrate, hydrate
Supplier: BeanTown Chemical
133905-100G
133905-500G
BT133905-100GEA
90.49
USD
BT133905-100G
BT133905-500G
Silicic acid hydrate, hydrate
Silicic acid hydrate
CAS: 1343-98-2; EC No: 215-683-2; MDL No: MFCD00054122; RTECS: VV8853000
-100 Mesh Powder; Linear Formula: SiO2·xH2O; MW: 60.09 (anhydrous)
- RTECS: VV8853000
- -100 Mesh Powder
Formula:
SiO₂·xH₂O MW: 60.09 g/mol (anhy) |
MDL Number:
MFCD00054122 CAS Number: 1343-98-2 EINECS: 215-683-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...