2-Chloronicotinic acid ≥99%

Supplier: BeanTown Chemical

Synonyms: 2-Chloro-3-pyridinecarboxylic acid, 2-Chloropyridine-3-carboxylic acid

133225-10G 133225-250G 133225-50G
BT133225-10GEA 24 USD
BT133225-10G BT133225-250G BT133225-50G
2-Chloronicotinic acid ≥99%
2-Chloronicotinic acid

CAS: 2942-59-8; EC No: 220-937-0; MDL No: MFCD00006236 Powder; Molecular Formula: C6H4ClNO2; MW: 157.55 Melting Point: 176-178°


  • Powder

Formula: C₆H₄ClNO₂
Melting Pt: 176…178 °C
MDL Number: MFCD00006236
CAS Number: 2942-59-8
EINECS: 220-937-0

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR