Bromocresol green ≥95% (dye content) ACS
Supplier: BeanTown Chemical
Synonyms:
3,3-Bis(3,5-dibromo-4-hydroxy-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
132970-25G
132970-5G
BT132970-25GEA
137.12
USD
BT132970-25G
BT132970-5G
Bromocresol green ≥95% (dye content) ACS
Bromocresol green
CAS: 76-60-8; EC No: 200-972-8; MDL No: MFCD00005874; RTECS: SJ7456000
Powder; Molecular Formula: C21H14Br4O5S; MW: 698.01
Melting Point: 225° (decomposes)
- RTECS: SJ7456000
- Powder
Formula:
C₂₁H₁₄Br₄O₅S MW: 698.02 g/mol Melting Pt: 225 °C (dec.) |
MDL Number:
MFCD00005874 CAS Number: 76-60-8 EINECS: 200-972-8 |
Specification Test Results
Clarity of solution | Passes test |
Visual transition interval (pH) | 3.8-5.4 |
Visual transition interval (color) | Greenish yellow-blue |
Loss on drying (110°C/1hr) | ≤3% |
Appearance | Yellow/brown powder |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...