2-Pyrrolidone ≥99%
Supplier: BeanTown Chemical
CAS: 616-45-5; EC No: 210-483-1; MDL No: MFCD00005270; RTECS: UY5715000
Liquid/Low Melting Solid; Molecular Formula: C4H7NO; MW: 85.10
Melting Point: 23-25°; Boiling Point: 245°; Flash point:
- RTECS: UY5715000
- Refractive index: 1.487
- Hygroscopic
- Liquid/Low Melting Solid
Formula:
C₄H₇NO Boiling Pt: 245 °C Melting Pt: 23…25 °C Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00005270 CAS Number: 616-45-5 EINECS: 210-483-1 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...