Bismuth ≥99.9999% (trace metals basis), rods Ø 10 mm. x 145 mm long
Supplier: BeanTown Chemical
132380-1PC
132380-5PC
BT132380-1PCEA
604
USD
BT132380-1PC
BT132380-5PC
Bismuth ≥99.9999% (trace metals basis), rods Ø 10 mm. x 145 mm long
Bismuth
CAS: 7440-69-9; EC No: 231-177-4; MDL No: MFCD00134033
Solid; Molecular Formula: Bi; MW: 208.98
Melting Point: 271°; Boiling Point: 1560°
Density (g/mL): 9.8
- Solid
Formula:
Bi Boiling Pt: 1560 °C Melting Pt: 271 °C Density: 9.8 g/mL |
MDL Number:
MFCD00134033 CAS Number: 7440-69-9 EINECS: 231-177-4 |
Specification Test Results
Assay | 99.9999% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...