Octylphosphonic acid ≥99%

Supplier: BeanTown Chemical

Synonyms: OPA, 1-Octylphosphonic acid, n-Octylphosphonic acid

132290-1G 132290-5G
BT132290-1GEA 75 USD
BT132290-1G BT132290-5G
Octylphosphonic acid ≥99%
Octylphosphonic acid

CAS: 4724-48-5; EC No: 225-218-5; MDL No: MFCD00015841 Crystalline; Molecular Formula: C8H19O3P; MW: 194.21 Melting Point: 93-98°


  • Crystalline

Formula: C₈H₁₉O₃P
MW: 194.21 g/mol
Melting Pt: 93…98 °C
MDL Number: MFCD00015841
CAS Number: 4724-48-5
EINECS: 225-218-5

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR