Polyethylene glycol 6,000

Supplier: BeanTown Chemical

Synonyms: PEG 6,000, PEG 6000

132095-1KG 132095-250G 132095-5KG
BT132095-1KGEA 80.87 USD
BT132095-1KG BT132095-250G BT132095-5KG
Polyethylene glycol 6,000
Polyethylene glycol 6,000

CAS: 25322-68-3; EC No: 203-473-3; MDL No: MFCD01779614; RTECS: TQ4100000 Powder/Flakes; Linear Formula: H(OCH2CH2)nOH; MW ≈6,000 Melting Point: 60-63° Density (g/mL): 1.2


  • RTECS: TQ4100000
  • Powder/Flakes

Formula: H(OCH₂CH₂)nOH
MW: 6000 g/mol
Melting Pt: 60…63 °C
Density: 1.2 g/mL
MDL Number: MFCD01779614
CAS Number: 25322-68-3
EINECS: 203-473-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR