Indium ≥99.995% (trace metals basis), wire Ø 2.0 mm

Supplier: BeanTown Chemical

131970-1M 131970-25CM 131970-5M
BT131970-1MEA 239 USD
BT131970-1M BT131970-25CM BT131970-5M
Indium ≥99.995% (trace metals basis), wire Ø 2.0 mm
Indium

CAS: 7440-74-6; EC No: 231-180-0; MDL No: MFCD00134048; RTECS: NL1050000 Solid; Molecular Formula: In; MW: 114.82 Melting Point: 156.6°; Boiling Point: 2072° Density (g/mL): 7.3


  • RTECS: NL1050000
  • Solid

Formula: In
MW: 114.82 g/mol
Boiling Pt: 2000
Melting Pt: 156
Density: 7.3 g/mL
MDL Number: MFCD00134048
CAS Number: 7440-74-6
EINECS: 231-180-0
Merck Index: 13,04971

Order Now

Specification Test Results

Assay 99.995%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR