Cerium(IV) oxide ≥99.99%

Supplier: BeanTown Chemical

Synonyms: Ceric oxide, Cerium dioxide

131630-100G 131630-25G 131630-500G
BT131630-100GEA 162 USD
BT131630-100G BT131630-25G BT131630-500G
Cerium(IV) oxide ≥99.99%
Cerium (IV) oxide

CAS: 1306-38-3; EC No: 215-150-4; MDL No: MFCD00010933; RTECS: FK6310000 14 Micron Powder; Molecular Formula: CeO2; MW: 172.12 Density (g/mL): 7.13


  • RTECS: FK6310000
  • 14 Micron Powder

Formula: CeO₂
Density: 7.13 g/mL
MDL Number: MFCD00010933
CAS Number: 1306-38-3
EINECS: 215-150-4

Order Now

Specification Test Results

Assay 99.99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR