Dimethyl-3-oxoglutarate ≥97%
Supplier: BeanTown Chemical
Synonyms:
Dimethyl 1,3-acetonedicarboxylate, Dimethyl acetone-1,3-dicarboxylate
131465-100G
131465-25G
131465-500G
BT131465-100GEA
91.58
USD
BT131465-100G
BT131465-25G
BT131465-500G
Dimethyl-3-oxoglutarate ≥97%
Dimethyl 3-oxoglutarate
CAS: 1830-54-2; EC No: 217-385-8; MDL No: MFCD00008462
Liquid; Molecular Formula: C7H10O5; MW: 174.16
Boiling Point: 237-241°; Flash point:
- Refractive index: 1.444
- Liquid
Formula:
C₇H₁₀O₅ Boiling Pt: 237…241 °C Density: 1.206 g/mL Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00008462 CAS Number: 1830-54-2 EINECS: 217-385-8 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...