2,4-Dichloronicotinc acid ≥97%
Supplier: BeanTown Chemical
Synonyms:
2,4-Dichloropyridine-3-carboxylic acid, 2,4-Dichloro-3-pyridinecarboxylic acid, 2,4-Dichloronicotinic acid
131455-1G
131455-250MG
BT131455-1GEA
124
USD
BT131455-1G
BT131455-250MG
2,4-Dichloronicotinc acid ≥97%
2,4-Dichloronicotinc acid
CAS: 262423-77-8; MDL No: MFCD06796292
Solid; Molecular Formula: C6H3Cl2NO2 ; MW: 192.00
Melting Point: 151-156°
- Solid
Formula:
C₆H₃Cl₂NO₂ Melting Pt: 151…156 °C |
MDL Number:
MFCD06796292 CAS Number: 262423-77-8 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...