Zinc ≥99.995% (trace metals basis), foil, Thickness 1.0 mm
Supplier: BeanTown Chemical
131415-100X100
131415-25X25MM
131415-50X50MM
BT131415-100X1EA
904
USD
BT131415-100X1
BT131415-25X25
BT131415-50X50
Zinc ≥99.995% (trace metals basis), foil, Thickness 1.0 mm
Zinc
CAS: 7440-66-6; EC No: 231-175-3; MDL No: MFCD00011291
Solid; Molecular Formula: Zn; MW: 65.39
Melting Point: 420°; Boiling Point: 907°
Density (g/mL): 7.133
- Solid
Formula:
Zn MW: 65.39 g/mol Boiling Pt: 908 Melting Pt: 420 Density: 7.133 g/mL |
MDL Number:
MFCD00011291 CAS Number: 7440-66-6 EINECS: 231-175-3 Merck Index: 12,10255 |
Specification Test Results
Assay | 99.995% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...