Carbon ≥99%, felt, Thickness 1.27 cm (0.5 in)

Supplier: BeanTown Chemical

Synonyms: Charcoal

130885-10X10CM 130885-30X30CM
BT130885-10X10EA 42.57 USD
BT130885-10X10 BT130885-30X30
Carbon ≥99%, felt, Thickness 1.27 cm (0.5 in)
Carbon

EC No: 231-955-3; MDL No: MFCD00133992 Solid; Molecular Formula: C; MW: 12.01


  • Solid

Formula: C
MW: 12.01 g/mol
Melting Pt: 3727
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-955-3
Merck Index: 13,01818

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR