Rhenium ≥99.99% (trace metals basis), slug Ø 12.7 mm
Supplier: BeanTown Chemical
130175-1EACH
130175-5EACH
BT130175-1EACHEA
467
USD
BT130175-1EACH
BT130175-5EACH
Rhenium ≥99.99% (trace metals basis), slug Ø 12.7 mm
Rhenium
CAS: 7440-15-5; EC No: 231-124-5; MDL No: MFCD00011195
Solid; Molecular Formula: Re; MW: 186.21
Melting Point: 3180°; Boiling Point: 5596°
Density (g/mL): 21.02
- Solid
Formula:
Re Boiling Pt: 5596 °C Melting Pt: 3180 °C Density: 21.02 g/mL |
MDL Number:
MFCD00011195 CAS Number: 7440-15-5 EINECS: 231-124-5 |
Specification Test Results
Assay | 99.99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...