1,10-Diaza-18-crown-6 (1,4,10,13-tetraoxa-7,16-diazacyclooctadecane kryptofix® 22) ≥96%
Supplier: BeanTown Chemical
Synonyms:
Diaza-18-crown-6, 1,4,10,13-Tetraoxa-7,16-diazacyclooctadecane
129465-1G
129465-250MG
BT129465-1GEA
217
USD
BT129465-1G
BT129465-250MG
1,10-Diaza-18-crown-6 (1,4,10,13-tetraoxa-7,16-diazacyclooctadecane kryptofix® 22) ≥96%
1,10-Diaza-18-crown-6 (1,4,10,13-Tetraoxa-7,16-diazacyclooctadecane, Kryptofix® 22)
CAS: 23978-55-4; EC No: 245-965-0; MDL No: MFCD00005112; RTECS: HM1650000
Crystalline/Powder; Molecular Formula: C12H26N2O4; MW: 262.35
Melting Point: 111-114°
- RTECS: HM1650000
- Crystalline/Powder
Formula:
C₁₂H₂₆N₂O₄ MW: 262.35 g/mol Melting Pt: 111…114 °C |
MDL Number:
MFCD00005112 CAS Number: 23978-55-4 EINECS: 245-965-0 |
Specification Test Results
Assay | 96% |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...