1,5-Diphenylcarbazide ACS

Supplier: BeanTown Chemical

Synonyms: Diphenylcarbazide, sym-Diphenylcarbazide, 1,5-Diphenylcarbohydrazide, 1,5-Diphenylcarbonohydrazide, N'',N'''-Diphenylcarbonohydrazide, sym. Diphenylcarbazide

129335-100G 129335-25G 129335-5G
BT129335-100GEA 147 USD
BT129335-100G BT129335-25G BT129335-5G
1,5-Diphenylcarbazide ACS
1,5-Diphenylcarbazide

CAS: 140-22-7; EC No: 205-403-7; MDL No: MFCD00003013; RTECS: FF2750000 Crystalline ; Molecular Formula: C13H14N4O; MW: 242.28 Melting Point: 170-175° Air Sensitive, Light Sensitive


  • RTECS: FF2750000
  • Air Sensitive, Light Sensitive
  • Crystalline

Formula: C₁₃H₁₄N₄O
MW: 242.28 g/mol
Boiling Pt: 454
Melting Pt: 170…175 °C
MDL Number: MFCD00003013
CAS Number: 140-22-7
EINECS: 205-403-7
Merck Index: 13,03353
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03cf8836a36f1077443ec2e3cfa151621925bd46&vid=a1b2bd876c3fbd9d6ded95c8430e4ec16bf5172b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR