(R)-(-)-Methyl-3-hydroxybutyrate ≥98%
Supplier: BeanTown Chemical
Synonyms:
(R)-Methyl 3-hydroxybutanoate, Methyl (R)-3-hydroxybutyrate, Methyl (R)-(-)-3-Hydroxybutyrate
129275-1G
129275-25G
129275-5G
BT129275-1GEA
31
USD
BT129275-1G
BT129275-25G
BT129275-5G
(R)-(-)-Methyl-3-hydroxybutyrate ≥98%
(R)-(-)-Methyl 3-hydroxybutyrate
CAS: 3976-69-0; EC No: 223-610-0; MDL No: MFCD00063289
Liquid; Molecular Formula: C5H10O3; MW: 118.13
Boiling Point: 56-58°/11 mmHg; Flash point: 76°C (163°F)
Density (g/mL): 1.055; Refractive Index: 1.421; Optical Rotation: [α]20/D -19.5°, neat
- Optical rotation: [α]20/D -19.5°, neat
- Refractive index: 1.421
- Liquid
Formula:
C₅H₁₀O₃ Boiling Pt: 56…58 °C/11 mmHg Density: 1.055 g/mL Flash Pt: 76 °C (163 °F) |
MDL Number:
MFCD00063289 CAS Number: 3976-69-0 EINECS: 223-610-0 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...