Lutetium(III) oxide ≥99.99%

Supplier: BeanTown Chemical

Synonyms: Lutetium oxide, Dilutetium trioxide

129195-1G 129195-25G 129195-5G
BT129195-1GEA 86 USD
BT129195-1G BT129195-25G BT129195-5G
Lutetium(III) oxide ≥99.99%
Lutetium (III) oxide

CAS: 12032-20-1; EC No: 234-764-3; MDL No: MFCD00011100 Powder; Molecular Formula: Lu2O3; MW: 397.94 Density (g/mL): 9.42 Hygroscopic


  • Hygroscopic
  • Powder

Formula: Lu₂O₃
Density: 9.42 g/mL
MDL Number: MFCD00011100
CAS Number: 12032-20-1
EINECS: 234-764-3

Order Now

Specification Test Results

Assay 99.99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR