Dowex® 1X2 (Cl) 100-200 (Cl), beads
Supplier: BeanTown Chemical
129070-1KG
129070-250G
129070-50G
BT129070-1KGEA
806
USD
BT129070-1KG
BT129070-250G
BT129070-50G
Dowex® 1X2 (Cl) 100-200 (Cl), beads
Dowex® 1X2 (Cl)
CAS: 69011-19-4; MDL No: MFCD00132715
Beads
MDL Number:
MFCD00132715 CAS Number: 9085-42-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...