Germanium ≥99.9999%, pieces ≤2 cm

Supplier: BeanTown Chemical

129040-100G 129040-25G 129040-5G
BT129040-100GEA 1346.95 USD
BT129040-100G BT129040-25G BT129040-5G
Germanium ≥99.9999%, pieces ≤2 cm
Germanium

CAS: 7440-56-4; EC No: 231-164-3; MDL No: MFCD00085310; RTECS: LY5200000 Solid, electrical resistivity: 40 ohm-cm ; Molecular Formula: Ge; MW: 72.64 Melting Point: 937°; Boiling Point: 2830° Density (g/mL): 5.35


  • RTECS: LY5200000
  • Solid, electrical resistivity: >40 ohm-cm

Formula: Ge
Boiling Pt: 2830 °C
Melting Pt: 937 °C
Density: 5.35 g/mL
MDL Number: MFCD00085310
CAS Number: 7440-56-4
EINECS: 231-164-3

Order Now

Specification Test Results

Assay 99.9999%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR