Triethanolamine (trolamine) ≥98%
Supplier: BeanTown Chemical
Synonyms:
Tris-(2-hydroxyethyl)amine, 2,2',2''-Nitrilotriethanol
128875-100G
128875-2.5KG
128875-500G
BT128875-100GEA
27.75
USD
BT128875-100G
BT128875-2.5KG
BT128875-500G
Triethanolamine (trolamine) ≥98%
Triethanolamine (Trolamine)
CAS: 102-71-6; EC No: 203-049-8; MDL No: MFCD00002855; RTECS: KL9275000
Liquid; Linear Formula: (HOCH2CH2)3N; Molecular Formula: C6H15NO3; MW: 149.19
Melting Point: 19-21°; Boiling Point: 190-193°/5 mmHg; Flash point: 185°C (365°F)
Density (g/mL): 1.124; Refractive Index: 1.485
Air Sensitive, Hygroscopic
- RTECS: KL9275000
- Refractive index: 1.485
- Air Sensitive, Hygroscopic
- Liquid
Formula:
C₆H₁₅NO₃ MW: 149.19 g/mol Boiling Pt: 190…193 °C/5 mmHg Melting Pt: 19…21 °C Density: 1.124 g/mL Flash Pt: 185 °C (365 °F) |
MDL Number:
MFCD00002855 CAS Number: 102-71-6 EINECS: 203-049-8 |
|
Specification Test Results
Assay | 98% |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...