2-Bromoisonicotinic acid ≥97%
Supplier: BeanTown Chemical
Synonyms:
2-Bromo-4-pyridinecarboxylic acid, 2-Bromopyridine-4-carboxylic acid
128190-1G
128190-5G
BT128190-1GEA
73
USD
BT128190-1G
BT128190-5G
2-Bromoisonicotinic acid ≥97%
2-Bromoisonicotinic acid
CAS: 66572-56-3; MDL No: MFCD01646069
Powder; Molecular Formula: C6H3BrNO2; MW: 202.01
Melting Point: 217-220°
- Powder
Formula:
C₆H₄BrNO₂ Melting Pt: 217…220 °C |
MDL Number:
MFCD01646069 CAS Number: 66572-56-3 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...