Zinc ≥99.8%, granules ACS -20 mesh

Supplier: BeanTown Chemical

128045-100G 128045-1KG 128045-500G
BT128045-100GEA 40.89 USD
BT128045-100G BT128045-1KG BT128045-500G
Zinc ≥99.8%, granules ACS -20 mesh
Zinc

CAS: 7440-66-6; EC No: 231-175-3; MDL No: MFCD00011291; RTECS: ZH1400000 Solid; Molecular Formula: Zn; MW: 65.39 Melting Point: 420°; Boiling Point: 907° Density (g/mL): 7.133


  • RTECS: ZH1400000
  • Solid

Formula: Zn
MW: 65.39 g/mol
Boiling Pt: 908
Melting Pt: 420
Density: 7.133 g/mL
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
Merck Index: 12,10255

Order Now

Specification Test Results

Assay 99.8% min.
Iron (Fe) ≤0.01%
Lead (Pb) ≤0.01%
Arsenic (As) ≤0.00001%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR