D-Cycloserine ≥98%

Supplier: BeanTown Chemical

128035-1G 128035-5G
BT128035-1GEA 55 USD
BT128035-1G BT128035-5G
D-Cycloserine ≥98%
D-Cycloserine

CAS: 68-41-7; EC No: 200-688-4; MDL No: MFCD00005353; RTECS: NY2975000 Powder; Molecular Formula: C3H6N2O2; MW: 102.09 Melting Point: 147° (decomposes) Optical Rotation: [α]20/D +115.0±5.0°, c = 2% in water Air Sensitive


  • RTECS: NY2975000
  • Optical rotation: [α]20/D +115.0±5.0°, c = 2% in water
  • Air Sensitive
  • Powder

Formula: C₃H₆N₂O₂
Melting Pt: 147 °C (dec.)
MDL Number: MFCD00005353
CAS Number: 68-41-7
EINECS: 200-688-4

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR