2',7'-Dichlorofluorescein
Supplier: BeanTown Chemical
Synonyms:
2',7'-Dichloro-3',6'-dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one
127805-25G
127805-5G
BT127805-25GEA
152
USD
BT127805-25G
BT127805-5G
2',7'-Dichlorofluorescein
2',7'-Dichlorofluorescein
CAS: 76-54-0; EC No: 200-968-6; MDL No: MFCD00005047
Solid; Molecular Formula: C20H10Cl2O5; MW: 401.20
Melting Point: 280° (decomposes)
- Solid
Formula:
C₂₀H₁₀Cl₂O₅ MW: 401.20 g/mol Melting Pt: 280 °C (dec.) |
MDL Number:
MFCD00005047 CAS Number: 76-54-0 EINECS: 200-968-6 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...