Bathophenanthroline ≥98%
Supplier: BeanTown Chemical
Synonyms:
4,7-Diphenyl-1,10-phenanthroline, (Bathophenanthroline)
127755-1G
127755-25G
127755-5G
BT127755-1GEA
71
USD
BT127755-1G
BT127755-25G
BT127755-5G
Bathophenanthroline ≥98%
Bathophenanthroline
CAS: 1662-01-7; EC No: 216-767-1; MDL No: MFCD00004976; RTECS: SF8427000
Crystalline; Molecular Formula: C24H16N2; MW: 332.40
Melting Point: 218-222°
- RTECS: SF8427000
- Crystalline
Formula:
C₂₄H₁₆N₂ Melting Pt: 218…222 °C |
MDL Number:
MFCD00004976 CAS Number: 1662-01-7 EINECS: 216-767-1 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...