1-Dodecansulfonic acid sodium salt ≥99%
Supplier: BeanTown Chemical
Synonyms:
Dodecane-1-sulfonic acid sodium salt, Sodium 1-dodecanesulphonate, Dodecane-1-sulphonic acid sodium salt, Sodium 1-dodecansulphonate, 1-Dodecanesulphonic acid, sodium salt, 1-Dodecanesulfonic acid, sodium salt, Sodium dodecane-1-sulphonate, Sodium dodecane-1-sulfonate, Sodium 1-dodecansulfonate, Sodium 1-dodecanesulfonate
127750-1G
127750-25G
127750-5G
BT127750-1GEA
37
USD
BT127750-1G
BT127750-25G
BT127750-5G
1-Dodecansulfonic acid sodium salt ≥99%
1-Dodecansulphonic acid sodium salt
CAS: 2386-53-0; EC No: 219-200-6; MDL No: MFCD00007527
Powder; Linear Formula: CH3(CH2)11SO3Na; MW: 272.39
Melting Point:
- Powder
Formula:
C₁₂H₂₅NaO₃S MW: 272.38 g/mol Melting Pt: >300 °C |
MDL Number:
MFCD00007527 CAS Number: 2386-53-0 EINECS: 219-200-6 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...