Ethyl-2-(triphenylphosphoranylidene)acetate ≥98%
Supplier: BeanTown Chemical
126625-10G
126625-250G
126625-50G
BT126625-10GEA
27
USD
BT126625-10G
BT126625-250G
BT126625-50G
Ethyl-2-(triphenylphosphoranylidene)acetate ≥98%
Ethyl-2-(triphenylphosphoranylidene)acetate
CAS: 1099-45-2; EC No: 214-151-7; MDL No: MFCD00009183
Crystalline; Linear Formula: CH3CH2O2CCH=P(C6H5)3; Molecular Formula: C22H21O2P; MW: 348.38
Melting Point: 126-130°
Air Sensitive
- Air Sensitive
- Crystalline
Formula:
C₂₂H₂₁O₂P Melting Pt: 126…130 °C |
MDL Number:
MFCD00009183 CAS Number: 1099-45-2 EINECS: 214-151-7 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...