Diethyl aminomalonate hydrochloride ≥98%

Supplier: BeanTown Chemical

Synonyms: Diethyl 2-aminomalonate hydrochloride

126570-10G 126570-250G 126570-50G
BT126570-10GEA 35 USD
BT126570-10G BT126570-250G BT126570-50G
Diethyl aminomalonate hydrochloride ≥98%
Diethyl aminomalonate hydrochloride

CAS: 13433-00-6; EC No: 236-556-8; MDL No: MFCD00012510 Crystalline; Molecular Formula: C7H13NO4·HCl; MW: 211.65 Melting Point: 165-170° (decomposes) Hygroscopic


  • Hygroscopic
  • Crystalline

Formula: C₇H₁₄ClNO₄
MW: 211.65 g/mol
Melting Pt: 165…170 °C (dec.)
MDL Number: MFCD00012510
CAS Number: 13433-00-6
EINECS: 236-556-8

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR