Platinum : rhodium (87:13) (w/w), wire 0.25 mm dia. for ISA type R standard grade thermocouple
Supplier: BeanTown Chemical
144550-100CM
144550-25CM
144550-500CM
BT144550-100CMEA
292
USD
BT144550-100CM
BT144550-25CM
BT144550-500CM
Platinum : rhodium (87:13) (w/w), wire 0.25 mm dia. for ISA type R standard grade thermocouple
Platinum : rhodium
MDL No: MFCD00198186
Solid; Formula: Pt:Rh; 87:13 wt%
- Solid