Sodium stearate

Supplier: BeanTown Chemical

Synonyms: Stearate sodium, Stearic acid sodium salt

135450-1KG 135450-250G 135450-5KG
BT135450-1KGEA 83.04 USD
BT135450-1KG BT135450-250G BT135450-5KG
Sodium stearate
Sodium stearate

CAS: 822-16-2; EC No: 212-490-5; MDL No: MFCD00036404 Powder; Linear Formula: CH3(CH2)16COONa; Molecular Formula: C18H35NaO2; MW: 306.46


  • Powder

Formula: C₁₈H₃₅NaO₂
MW: 306.46 g/mol
Melting Pt: 288
MDL Number: MFCD00036404
CAS Number: 822-16-2
EINECS: 212-490-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR