3-Bromo-p-toluic acid ≥98%

Supplier: BeanTown Chemical

126520-100G 126520-25G 126520-5G
BT126520-100GEA 392 USD
BT126520-100G BT126520-25G BT126520-5G
3-Bromo-p-toluic acid ≥98%
3-Bromo-p-toluic acid

CAS: 7697-26-9; EC No: 231-712-1; MDL No: MFCD00002488 Solid; Molecular Formula: C8H7BrO2; MW: 215.04


  • Solid

Formula: C₈H₇BrO₂
MDL Number: MFCD00002488
CAS Number: 7697-26-9
EINECS: 231-712-1
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR