n-Pentacosane ≥99%
Supplier: BeanTown Chemical
Synonyms:
N-pentacosane
126375-1G
126375-25G
126375-5G
BT126375-1GEA
58
USD
BT126375-1G
BT126375-25G
BT126375-5G
n-Pentacosane ≥99%
n-Pentacosane
CAS: 629-99-2; EC No: 211-123-6; MDL No: MFCD00009353
Solid; Linear Formula: CH3(CH2)23CH3; Molecular Formula: C25H52; MW: 352.67
Melting Point: 53-56°; Boiling Point: 169-170°/0.05 mmHg; Flash point: 120°C (248°F)
Density (g/mL): 0.779; Refractive Index: 1.438
- Refractive index: 1.438
- Solid
Formula:
H₃C(CH₂)₂₃CH₃ MW: 352.69 g/mol Boiling Pt: 169…170 °C/0.05 mmHg Melting Pt: 53…56 °C Density: 0.779 g/mL Flash Pt: 120 °C (248 °F) |
MDL Number:
MFCD00009353 CAS Number: 629-99-2 EINECS: 211-123-6 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...