Sodium carbonate ≥99.5% ACS
Supplier: BeanTown Chemical
Synonyms:
Soda, Carbonic acid sodium salt, Calcined soda, Disodium carbonate, Carbonic acid disodium salt, Na-X, Bisodium carbonate, Carbonic acid, disodium salt, Soda ash
125885-2KG
125885-500G
BT125885-2KGEA
89.01
USD
BT125885-2KG
BT125885-500G
Sodium carbonate ≥99.5% ACS
Sodium carbonate
CAS: 497-19-8; EC No: 207-838-8; MDL No: MFCD00003494; RTECS: VZ4050000
Powder; Linear Formula: Na2CO3; MW: 105.99
Melting Point: 851°
Density (g/mL): 2.532; Refractive Index: 1.535
Hygroscopic
- RTECS: VZ4050000
- Refractive index: 1.535
- Hygroscopic
- Powder
Formula:
Na₂CO₃ MW: 105.99 g/mol Boiling Pt: 1600 Melting Pt: 854 Density: 2.532 g/mL |
MDL Number:
MFCD00003494 CAS Number: 497-19-8 EINECS: 207-838-8 Merck Index: 14,08596 |
|
Specification Test Results
Assay | 99.5% min. |
Loss on heating (285°C) | ≤1.0% |
Insoluble matter | ≤0.01% |
Nitrogen compounds (as N) | ≤0.001% |
Chloride (Cl) | ≤0.001% |
Phosphate (PO4) | ≤0.001% |
Heavy metals (as Pb) | ≤5ppm |
Ammonium hydroxide PPT | ≤0.01% |
Iron (Fe) | ≤5ppm |
Arsenic (As) | ≤1ppm |
Potassium (K) | ≤0.005% |
Sulfur compounds | ≤0.003% |
Silica (SiO2) | ≤0.005% |
Calcium by AA (Ca) | ≤0.02% |
Magnesium (Mg) | ≤0.004% |
Clarity of solution | Clear and complete |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...