Glycerol monostearate

Supplier: BeanTown Chemical

Synonyms: Glyceryl monostearate

125575-1KG 125575-250G 125575-5KG
BT125575-1KGEA 86 USD
BT125575-1KG BT125575-250G BT125575-5KG
Glycerol monostearate
Glycerol monostearate

CAS: 31566-31-1; EC No: 250-705-4; MDL No: MFCD00036186 Powder; Molecular Formula: C21H42O4; MW: 358.56


  • Powder

Formula: C₂₁H₄₄O₅
MW: 376.57 g/mol
MDL Number: MFCD00036186
CAS Number: 31566-31-1
EINECS: 250-705-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR