Lithium carbonate ≥99%

Supplier: BeanTown Chemical

125545-2KG 125545-500G
BT125545-2KGEA 863.76 USD
BT125545-2KG BT125545-500G
Lithium carbonate ≥99%
Lithium carbonate

CAS: 554-13-2; EC No: 209-062-5; MDL No: MFCD00011084 Powder; Linear Formula: Li2CO3; MW: 73.90 Melting Point: 618°


  • Powder

Formula: Li₂CO₃
MW: 73.89 g/mol
Melting Pt: 720
MDL Number: MFCD00011084
CAS Number: 554-13-2
EINECS: 209-062-5
Merck Index: 13,05549

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR