Sodium 1-undecanesulfonate ≥98% (dry weight)

Supplier: BeanTown Chemical

125260-1G 125260-25G 125260-5G
BT125260-1GEA 16 USD
BT125260-1G BT125260-25G BT125260-5G
Sodium 1-undecanesulfonate ≥98% (dry weight)
Sodium 1-undecanesulphonate

CAS: 5838-34-6; EC No: 000-000-0; MDL No: MFCD00053627 Powder; Linear Formula: CH3(CH2)10SO3Na; MW: 258.36 Melting Point: 300°


  • Powder

Formula: C₁₂H₂₅NaO₃S
MW: 258.36 g/mol
Melting Pt: >300 °C
MDL Number: MFCD00053627
CAS Number: 5838-34-6

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR