Strontium fluoride ≥99.99%
Supplier: BeanTown Chemical
125040-25G
125040-500MG
125040-5G
BT125040-25GEA
208.79
USD
BT125040-25G
BT125040-500MG
BT125040-5G
Strontium fluoride ≥99.99%
Strontium fluoride
CAS: 7783-48-4; EC No: 232-000-3; MDL No: MFCD00011251; RTECS: WK8925000
Powder; Linear Formula: SrF2; MW: 125.62
Melting Point: 1473°; Boiling Point: 2489°
Density (g/mL): 4.24; Refractive Index: 1.442
- RTECS: WK8925000
- Refractive index: 1.442
- Powder
Formula:
F₂Sr Boiling Pt: 2489 °C Melting Pt: 1473 °C Density: 4.24 g/mL |
MDL Number:
MFCD00011251 CAS Number: 7783-48-4 EINECS: 232-000-3 UN: 3288 ADR: 6.1,III |
Specification Test Results
Assay | 99.99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...