Potassium carbonate ≥99.995%

Supplier: BeanTown Chemical

Synonyms: Potash, Carbonic acid potassium salt, Carbonic acid, dipotassium salt

124785-100G 124785-25G 124785-5G
BT124785-100GEA 264 USD
BT124785-100G BT124785-25G BT124785-5G
Potassium carbonate ≥99.995%
Potassium carbonate

CAS: 584-08-7; EC No: 209-529-3; MDL No: MFCD00011382; RTECS: TS7750000 Powder; Linear Formula: K2CO3; MW: 138.21 Melting Point: 891° Density (g/mL): 2.43 Hygroscopic


  • RTECS: TS7750000
  • Hygroscopic
  • Powder

Formula: K₂CO₃
MW: 138.21 g/mol
Melting Pt: 891
Density: 2.43 g/mL
MDL Number: MFCD00011382
CAS Number: 584-08-7
EINECS: 209-529-3
Merck Index: 13,07702
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d5900faa1ba52c3213eed8f6bb2d952bac1f30f&vid=6298361525e6af416556b5694c74a8a91faa437c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.995%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR