Iron(II) sulfide ≥99.9%
Supplier: BeanTown Chemical
Synonyms:
Ferrous sulphide, Ferrous sulfide
124300-10G
124300-50G
BT124300-10GEA
55
USD
BT124300-10G
BT124300-50G
Iron(II) sulfide ≥99.9%
Iron (II) sulphide
CAS: 1317-37-9; EC No: 215-268-6; MDL No: MFCD00011013
-100 Mesh Powder; Molecular Formula: FeS; MW: 87.91
Density (g/mL): 4.74
Air Sensitive, Moisture Sensitive
- Air Sensitive, Moisture Sensitive
- -100 Mesh Powder
Formula:
FeS MW: 87.91 g/mol Melting Pt: 1195 Density: 4.74 g/mL |
MDL Number:
MFCD00011013 CAS Number: 1317-37-9 EINECS: 215-268-6 UN: 3077 ADR: 9 III Merck Index: 12,04106 |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...